Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene-3-carboxylic acid, 99%

Catalog No. p-7022797
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 88-13-1 | C5H4O2S | 128.145 g/mol

3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiophenecarboxylic acid has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a lading compound for the development of a clinic useful D-amino acid inhibitor and have the potential to sever as active site proves to elucidate the structure-function relationships of D-amino acids.

Solubility
Soluble in water at 20°C 4.3 g/L.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-13-1
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.145
MDL Number MFCD00005467
InChI Key YNVOMSDITJMNET-UHFFFAOYSA-N
Synonym 3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be
PubChem CID 6918
IUPAC Name thiophene-3-carboxylic acid
SMILES C1=CSC=C1C(=O)O

Specifications

Melting Point 137°C to 141°C
Quantity 5 g
Beilstein 1994
Merck Index 14,9279
Solubility Information Soluble in water at 20°C 4.3g/L.
Formula Weight 128.15
Percent Purity 99%
Chemical Name or Material Thiophene-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-802-5
RTECSNumber XM8330300
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.