Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetonitrile, 98%

Catalog No. AC139375000
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 545-06-2 | C2Cl3N | 144.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545-06-2
Molecular Formula C2Cl3N
Molecular Weight (g/mol) 144.39
InChI Key DRUIESSIVFYOMK-UHFFFAOYSA-N
Synonym trichloroacetonitrile, acetonitrile, trichloro, tritox, cyanotrichloromethane, trichloromethyl cyanide, trichloroethanenitrile, trichlouracetonitril, trichlor-acetonitrile, trichloromethylnitrile, nitrile trichloracetique
PubChem CID 11011
ChEBI CHEBI:82541
IUPAC Name 2,2,2-trichloroacetonitrile
SMILES C(#N)C(Cl)(Cl)Cl

Specifications

Melting Point -42.0°C
Color Colorless to Yellow
Density 1.4400g/mL
Boiling Point 83.0°C to 84.0°C
Flash Point 195°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Linear Formula CCl3CN
Quantity 500 g
Beilstein 02,212
Fieser 01,1194; 05,686; 06,604; 07,381; 12,526; 13,32
Specific Gravity 1.44
Merck Index 15,9793
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Formula Weight 144.39
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trichloroacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 208-885-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.