Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophene, 99%

Catalog No. AAA1394130
Click to view available options
Quantity:
50 g
250 g
500 g

CAS: 110-02-1 | C4H4S | 84.14 g/mol

Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene is an important building block in dyes, agrochemicals and pharmaceuticals synthesis. It is involved in the chloroalkylation reactions in 2,5-positions. It is also used to prepare butane by reduction with raney nickel, 2-vinylthiophene, dithienyl, and 2-halo thiophenes by reacting with halogens.

Solubility
Miscible with ethyl alcohol, diethyl ether and other organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-02-1
Molecular Formula C4H4S
Molecular Weight (g/mol) 84.14
MDL Number MFCD00005413
InChI Key YTPLMLYBLZKORZ-UHFFFAOYSA-N
Synonym thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
PubChem CID 8030
ChEBI CHEBI:30856
IUPAC Name thiophene
SMILES S1C=CC=C1

Specifications

Melting Point -38°C
Density 1.066
Boiling Point 83°C to 85°C
Flash Point −9°C (16°F)
Odor Stench
Refractive Index 1.528
Quantity 250 g
UN Number UN2414
Beilstein 103222
Merck Index 14,9353
Solubility Information Miscible with ethyl alcohol,diethyl ether and other organic solvents. Immiscible with water.
Formula Weight 84.14
Percent Purity 99%
Chemical Name or Material Thiophene
Show More Show Less
Hazard Category H225-H302-H319
Hazard Statement GHS H Statement
H225-H331-H302-H312-H315-H319
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: THIOPHENE
EINECSNumber 203-729-4
RTECSNumber XM7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.