Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiophosphoryl-PmmH-6 Dendrimer, Generation 1.5, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
169132-80-3
C66H54N6O15P4S4
1423.32
MFCD03458724
GVUPCQKZNKROEB-UHFFFAOYSA-N
thiophosphoryl-pmmh-6 dendrimer, generation 1.5
131874880
CN(N=CC1=CC=C(C=C1)OP(=S)(OC2=CC=C(C=C2)C=NN(C)P(=S)(OC3=CC=C(C=C3)C=O)OC4=CC=C(C=C4)C=O)OC5=CC=C(C=C5)C=NN(C)P(=S)(OC6=CC=C(C=C6)C=O)OC7=CC=C(C=C7)C=O)P(=S)(OC8=CC=C(C=C8)C=O)OC9=CC=C(C=C9)C=O
169132-80-3
C66H54N6O15P4S4
1423.32
MFCD03458724
GVUPCQKZNKROEB-UHFFFAOYSA-N
thiophosphoryl-pmmh-6 dendrimer, generation 1.5
131874880
CN(N=CC1=CC=C(C=C1)OP(=S)(OC2=CC=C(C=C2)C=NN(C)P(=S)(OC3=CC=C(C=C3)C=O)OC4=CC=C(C=C4)C=O)OC5=CC=C(C=C5)C=NN(C)P(=S)(OC6=CC=C(C=C6)C=O)OC7=CC=C(C=C7)C=O)P(=S)(OC8=CC=C(C=C8)C=O)OC9=CC=C(C=C9)C=O
Packaging Glass bottle
Quantity 500mg
NMR Complies
Infrared Spectrum Authentic
Color Cream to White
Formula Weight 1423.35
Physical Form Powder
Chemical Name or Material Thiophosphoryl-PMMH-6 Dendrimer, Generation 1.5, Generation 1.5