Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-1 Antibody (092), FITC

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290731F
Description
Thioredoxin-1 Monoclonal specifically detects Thioredoxin-1 in Mouse samples. It is validated for ELISA.Specifications
| Thioredoxin-1 | |
| Monoclonal | |
| FITC | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 92 | |
| PBS | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Thioredoxin-1 (P10639; Met1-Val105). | |
| 0.1 mL | |
| Breast Cancer, Lipid and Metabolism | |
| 7295 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction