Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-1 Antibody (116), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289458G
Description
Thioredoxin-1 Monoclonal specifically detects Thioredoxin-1 in Human samples. It is validated for Western Blot.Specifications
Thioredoxin-1 | |
Monoclonal | |
DyLight 488 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
116 | |
50 mM Sodium Borate | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Thioredoxin-1 (P10599; Met1-Val105). | |
0.1 mL | |
Breast Cancer, Lipid and Metabolism | |
7295 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction