Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP252983MFV450
Description
Thioredoxin-1 Polyclonal antibody specifically detects Thioredoxin-1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Thioredoxin-1 | |
Polyclonal | |
Flow Cytometry | |
TRDX;TRX;TRX1;Trx80;TXN | |
Full length recombinant human Thioredoxin protein (amino acids 1-105) [UniProt P10599] | |
0.1 mL | |
Breast Cancer, Lipid and Metabolism | |
7295 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction