Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioredoxin 1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA590044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590044 Supplier Thermo Scientific Supplier No. PA590044

Rabbit Polyclonal Antibody

Immunogen sequence: VKQIESKTAF QEALDAAGDK LVVVDFSATW CGPCKMIKPF FHSLSEKYSN VIFLEVDVDD CQDVASECEV KCMPTFQFFK KGQKVGEFSG ANKEKLEATI NELV

Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cysteine residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds. Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations. Specific protein disulfide targets for reduction by Trx include protein disulfide-isomerase (PDI) and a number of transcription factors such as p53, NF-kB and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thioredoxin 1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene TXN
Gene Accession No. P10599, P10639, P11232
Gene Alias ADF, ATL-derived factor, AW550880, DKFZp686B1993, MGC61975, RP11-427L11.1, SASP, surface-associated sulphydryl protein, testicular tissue protein Li 199, thioredoxin, thioredoxin 1, thioredoxin delta 3, TRDX, TRX, TRX1, Txn, TXN delta 3, Txn1
Gene Symbols TXN, Txn1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 2-105 of human Trx1/Thioredoxin 1 (Trx1/TXN) (NP_0033202)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116484, 22166, 7295
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.