Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-2 Antibody (054), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290123F
Description
Thioredoxin-2 Monoclonal specifically detects Thioredoxin-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Thioredoxin-2 | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
54 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
mitochondrial thioredoxin, MTRX, MT-TRX, thioredoxin 2, thioredoxin, mitochondrial, thioredoxin-2, TRX2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Thioredoxin-2 (Q99757; Thr60-Gly166). | |
0.1 mL | |
Breast Cancer | |
25828 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction