Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-2 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300013PE
Description
Thioredoxin-2 Polyclonal antibody specifically detects Thioredoxin-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Thioredoxin-2 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
mitochondrial thioredoxin, MTRX, MT-TRX, thioredoxin 2, thioredoxin, mitochondrial, thioredoxin-2, TRX2 | |
Produced in rabbits immunized with purified, recombinant Human Thioredoxin-2 (Uniprot#: Q99757; Thr60-Gly166) | |
0.1 mL | |
Breast Cancer | |
25828 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction