Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin-like 5/TRP14/TXNDC17 Antibody (043), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289879MFV610
Description
Thioredoxin-like 5/TRP14/TXNDC17 Monoclonal antibody specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Human samples. It is validated for ELISASpecifications
Thioredoxin-like 5/TRP14/TXNDC17 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
84817 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
043 | |
ELISA | |
MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, thioredoxin-like 5, Thioredoxin-like protein 5, TRP1414 kDa thioredoxin-related protein, TXNL5 | |
Rabbit immunized with purified, recombinant Human Thioredoxin-like 5/TRP14/TXNDC17 (Uniprot#: Q9BRA2, Met1-Asp123) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction