Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioredoxin Reductase 1/TRXR1 Antibody (30F1), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259489DL594
This item is not returnable.
View return policy
Description
Thioredoxin Reductase 1/TRXR1 Monoclonal antibody specifically detects Thioredoxin Reductase 1/TRXR1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Thioredoxin Reductase 1/TRXR1 | |
Monoclonal | |
DyLight 594 | |
EC 1.8.1, EC 1.8.1.9, Gene associated with retinoic and IFN-induced mortality 12 protein, Gene associated with retinoic and interferon-induced mortality 12 protein, GRIM12, GRIM-12TR1, KDRF, KM-102-derived reductase-like factor, MGC9145, oxidoreductase, thioredoxin reductase 1, thioredoxin reductase 1, cytoplasmic, thioredoxin reductase GRIM-12, Thioredoxin reductase TR1, TR, Trxr1, TXNRTRXR1 | |
Recombinant human Thioredoxin Reductase 1/TRXR1 (161-647aa) purified from E. coli | |
0.1 mL | |
Breast Cancer, Lipid and Metabolism | |
7296 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunofluorescence | |
30F1 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction