Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thiosalicylic acid

Catalog No. T33200500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33200500G 500 g
T33200100G 100 g
T332005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T33200500G Supplier Sigma Aldrich Supplier No. T33200500G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thiosalicylic acid

Specifications

CAS 147-93-3
Melting Point 162°C to 165°C (lit.)
Molecular Formula C7H6O2S
Linear Formula HSC6H4CO2H
MDL Number MFCD00004836
Synonym 2-Mercaptobenzoic acid
Molecular Weight (g/mol) 154.19
Percent Purity 97%
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.