Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thioster-containing protein 1 Rabbit anti-Insect, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0632R

Description
Telomerase is an RNA-dependent DNA polymerase that uses an RNA component to add telomeric repeat sequences at the ends of chromosomes. Besides the RNA component which serves as the template that specifies the telomeric repeat, the telomerase complex contains a reverse transcriptase protein (TRT) and various accessory proteins including the telomerase-associated protein 1 (TP1). Telomerase activity is low in most somatic cells, causing the gradual shortening of telomeres which can ultimately lead to telomere fusion and cell death. High levels of telomerase activity are widely seen in cancerous cells and while recent experiments have suggested that telomerase may be a viable target in cancer therapy, expression levels of TP1 do not correlate with malignancy. At least two isoforms of TP1 are known to exist.Specifications
Thioster-containing protein 1 | |
Polyclonal | |
Unconjugated | |
p230; p240; p80 homolog; p80 telomerase homolog; rTLP1; telomerase associated protein 1; Telomerase protein 1; Telomerase protein component 1; Telomerase-associated protein 1; Tep1; TLP1; TP1; TROVE domain family, member 1; TROVE1; VAULT2 | |
KLH conjugated synthetic peptide derived from Anopheles stephensi Thioster-containing protein 1 | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
Antibody with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Rabbit | |
Protein A | |
RUO | |
Insect | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction