Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiostrepton, G-Biosciences
SCI_ED

Catalog No. S000017
Click to view available options
Quantity:
1 g
5 g

Thiostrepton is a cyclic peptide antibiotic isolated from Streptomyces azureus. It targets a wide variety of gram-negative and gram-positive bacteria, especially those causing skin infections. It has been used as a marker for selection of recombinant Streptomyces cultures.

Thiostrepton has been shown to inhibit FOXM1, a gene commonly overexpressed in cancer cell lines, thus inhibiting cancer cell growth. Thiostrepton functions by binding elongation factor G (EF-G) and interfering with ribosomal GTPase activity, ultimately inhibiting protein synthesis.

Chemical Identifiers

Molecular Weight (g/mol) 1664.89 g/mol

Specifications

Quantity 5 g
Percent Purity 95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.