Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiourea, 99%

Catalog No. p-7020764
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g

CAS: 62-56-6 | CH4N2S | 76.12 g/mol

Reagent for organic synthesisUsed as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It acts as a catalyst for asymmetric reactions. It plays an essential role as a catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for organic synthesisUsed as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It acts as a catalyst for asymmetric reactions. It plays an essential role as a catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.

Solubility
Soluble in water.

Notes
Stable. Incompatible with strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-56-6
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00008067
InChI Key UMGDCJDMYOKAJW-UHFFFAOYSA-N
Synonym thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
PubChem CID 2723790
ChEBI CHEBI:36946
IUPAC Name thiourea
SMILES NC(N)=S

Specifications

Melting Point 172°C to 178°C
Density 1.405
Odor Odorless
Quantity 100 g
UN Number UN3077
Beilstein 605327
Merck Index 14,9367
Solubility Information Soluble in water.
Formula Weight 76.12
Percent Purity 99%
Chemical Name or Material Thiourea
Show More Show Less
Hazard Category H302-H350-H361d-H500
Hazard Statement GHS H Statement
H351-H361-H302
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-543-5
RTECSNumber YU2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.