Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioxanthene, 98%

Catalog No. p-3598499
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 261-31-4 | C13H10S | 198.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261-31-4
Molecular Formula C13H10S
Molecular Weight (g/mol) 198.28
MDL Number MFCD00046936
InChI Key PQJUJGAVDBINPI-UHFFFAOYSA-N
Synonym thioxanthene, 10h-dibenzo b,e thiin, thiaxanthen, thioxanthen, dibenzothiapyran, 9h-thioxanthen, 10h-dibenzo b,e thiopyran, chembl79451
PubChem CID 67495
ChEBI CHEBI:51055
IUPAC Name 9H-thioxanthene
SMILES C1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 128°C to 131°C
Color White
Boiling Point 340°C (730.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 9522
Solubility Information Solubility in water: .. Other solubilities: freely soluble in chloroform,moderately soluble in alcohol,ether
Formula Weight 198.29
Percent Purity 98%
Physical Form Crystals or Needles
Chemical Name or Material Thioxanthene
Show More Show Less
EINECSNumber 205-972-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.