Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THNSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575923 Supplier Invitrogen™ Supplier No. PA575923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

THNSL2 (threonine synthase-like 2), also known as TSH2, is a 484 amino acid protein belonging to the threonine synthase family. Utilizing pyridoxal phosphate as a cofactor, THNSL2 may function as a catabolic phospholyase on gamma and beta phosphorylated substrates. THNSL2 may also degrade O-phospho-threonine to alpha-ketobutyrate, ammonia and phosphate. Existing as four alternatively spliced isoforms, THNSL2 is encoded by a gene mapping to human chromosome 2p11.2. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8 gene defects. An extremely rare recessive genetic disorder, Alstrom syndrome, is related to mutations in the ALMS1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THNSL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene THNSL2
Gene Accession No. Q5M7T9, Q80W22, Q86YJ6
Gene Alias BC051244; FLJ10916; FLJ35504; RGD1309144; secreted osteoclastogenic factor of activated T cells; Secreted osteoclastogenic factor of activated T-cells; SOFAT; THNSL2; threonine synthase like 2; threonine synthase-like 2; threonine synthase-like 2 (bacterial); threonine synthase-like 2 (S. cerevisiae); THS2; TSH2
Gene Symbols THNSL2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 103-148 in Human THNSL2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232078, 297332, 55258
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.