Learn More
THOC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302839AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
P84/N5-5E10 recognizes an 84 kDa nuclear matrix protein. Initially isolated by its interaction with the amino terminal region of the retinoblastoma protein, it has been used extensively as a nuclear-specific marker in both immunofluorescence and cell fractionation studies.Specifications
THOC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96FV9 | |
THOC1 | |
Between 607 and 657 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
THOC1 | |
HPR1, P84, P84N5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9984 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.