Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THOC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561623
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561623 Supplier Invitrogen™ Supplier No. PA561623
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHKWHYKLTK ASVHCLETGE YTHIRNILIV LTKILPWYPK VLNLGQALER RVHKICQEEK EKRPDLYALA MGYSGQLKSR KSYMIPENEF HHKDPPPRNA VASV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

The TREX multiprotein complex binds specifically to spliced mRNAs to facilitate mRNA export. The protein encoded by this gene is a member of the THO complex, a subset of the TREX complex. The encoded protein interacts with the THOC1 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THOC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THOC2
Gene Accession No. Q8NI27
Gene Alias 6330441O12Rik; CXorf3; D130005M13Rik; dJ506G2.1; Gm1139; Gm1793; hTREX120; mental retardation, X-linked 12; mental retardation, X-linked 35; MRX12; MRX35; RGD1561623; THO complex 2; THO complex subunit 2; Tho2; THOC2; transcription/export complex subunit 2
Gene Symbols THOC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THOC2. Recombinant protein control fragment (Product #RP-100593).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.