Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THOC2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984001
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4001 Supplier Bioss Supplier No. BS1663R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The TREX multiprotein complex binds specifically to spliced mRNAs to facilitate mRNA export. The protein encoded by this gene is a member of the THO complex, a subset of the TREX complex. The encoded protein interacts with the THOC1 protein.

Specifications

Antigen THOC2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene THOC2
Gene Accession No. Q8NI27
Gene Alias 6330441O12Rik; CXorf3; D130005M13Rik; dJ506G2.1; Gm1139; Gm1793; hTREX120; mental retardation, X-linked 12; mental retardation, X-linked 35; MRX12; MRX35; THO complex 2; THO complex subunit 2; Tho2; Thoc2; transcription/export complex subunit 2
Gene Symbols THOC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human THOC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57187
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.