Learn More
Invitrogen™ THOC3 Polyclonal Antibody

Description
Immunogen sequence: KTHRSKAEEQ FKFEVNEISW NNDNNMFFLT NGNGCINILS YPELKPVQSI NAHPSNCICI KFDPMGKYFA TGSADALV Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | THOC3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | THOC3 |
| Gene Accession No. | Q96J01 |
| Gene Alias | 2410044K02Rik; AL033344; hTREX45; LOC529231 protein; TEX1; TEX1 homolog; THO complex 3; THO complex subunit 3; Tho3; Thoc3; zgc:100815 |
| Gene Symbols | THOC3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.