Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THOC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560586 Supplier Invitrogen™ Supplier No. PA560586
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HDGKMLASAS EDHFIDIAEV ETGDKLWEVQ CESPTFTVAW HPKRPLLAFA CDDKDGKYDS SREAGTVKLF GLPNDS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THOC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THOC3
Gene Accession No. Q8VE80, Q96J01
Gene Alias 2410044K02Rik; AL033344; hTREX45; LOC529231 protein; TEX1; TEX1 homolog; THO complex 3; THO complex subunit 3; Tho3; Thoc3; zgc:100815
Gene Symbols THOC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THOC3. Recombinant protein control fragment (Product #RP-99038).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290519, 73666, 84321
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.