Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THOC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | THOC3 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111161
|
Invitrogen™
PA5111161 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111210
|
Invitrogen™
PA5111210 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: PSNPDLFVTA SGDKTIRIWD VRTTKCIATV NTKGENINIC WSPDGQTIAV GNKDDVVT.
THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.Specifications
THOC3 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96J01 | |
84321 | |
Recombinant Protein corresonding to Human THOC3. Recombinant protein control fragment (Product #RP-99037). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
THOC3 | |
2410044K02Rik; AL033344; hTREX45; LOC529231 protein; TEX1; TEX1 homolog; THO complex 3; THO complex subunit 3; Tho3; Thoc3; zgc:100815 | |
THOC3 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title