Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THOC5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | THOC5 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
THOC5 Polyclonal specifically detects THOC5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
THOC5 | |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
8563 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SDAEEEQTTKRRRPTLGVQLDDKRKEMLKRHPLSVMLDLKCKDDSVLHLTFYYLMNLNIMTVKAKVTTAMELITPISAGDLLSPDSVLSCLYPGDHGKKTPNPANQYQFDKVGILTLSDYVLELGHPYLWVQKLGG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C22orf19, chromosome 22 open reading frame 19, Fmip, Fms-interacting protein, fSAP79hTREX90, Functional spliceosome-associated protein 79, KIAA0983, NF2/meningioma region protein pK1.3, PK1.3, Placental protein 39.2, PP39.2, THO complex 5, THO complex subunit 5 homolog | |
THOC5 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title