Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THOC5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP119160
Description
THOC5 Polyclonal specifically detects THOC5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
THOC5 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
C22orf19, chromosome 22 open reading frame 19, Fmip, Fms-interacting protein, fSAP79hTREX90, Functional spliceosome-associated protein 79, KIAA0983, NF2/meningioma region protein pK1.3, PK1.3, Placental protein 39.2, PP39.2, THO complex 5, THO complex subunit 5 homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
8563 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
NP_003669.4 | |
THOC5 | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human THO complex 5 using the numbering given in entry NP_003669.4 (GeneID 8563). | |
0.1 mL | |
Primary | |
Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, European domestic ferret, African elephant, Chinese hamster and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction