Learn More
Invitrogen™ THOC5 Polyclonal Antibody

Description
Positive Control: NIH/3T3 cell lysate, PC-12 cell lysate, Hela cell lysate, HepG2 cell lysate, human ovarian tumor tissue, mouse brain tissue. Subcellular Location: Nucleus, Cytoplasm.
Specifications
Specifications
| Antigen | THOC5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | THOC5 |
| Gene Accession No. | Q13769, Q68FX7, Q8BKT7 |
| Gene Alias | 1700060C24Rik; A430085L24Rik; C22orf19; Fmip; Fms interacting protein; Fms-interacting protein; fSAP79; Functional spliceosome-associated protein 79; hTREX90; KIAA0983; NF2/meningioma region protein pK1.3; PK1.3; placental protein 39.2; PP39.2; serum/glucocorticoid regulated kinase 2; Sgk2; THO complex 5; THO complex subunit 5 homolog; Thoc5 |
| Gene Symbols | THOC5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.