Learn More
THOC5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | THOC5 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-1822
![]() |
Bethyl Laboratories, Inc
A302119AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Component the THO subcomplex of the TREX complex. The TREX complex specifically associates with spliced mRNA and not with unspliced pre-mRNA. It is recruited to spliced mRNAs by a transcription-independent mechanism. Binds to mRNA upstream of the exon-junction complex (EJC) and is recruited in a splicing- and cap-dependent manner to a region near the 5' end of the mRNA where it functions in mRNA export. The recruitment occurs via an interaction between THOC4 and the cap-binding protein NCBP1. The TREX complex is essential for the export of Kaposi's sarcoma-associated herpesvirus (KSHV) intronless mRNAs and infectious virus production. The recruitment of the TREX complex to the intronless viral mRNA occurs via an interaction beteen KSHV ORF57 protein and THOC4. UAP56 functions as a bridge between THOC4 and the THO complex. THOC5 in conjunction with THOC4 functions in NXF1-NXT1 mediated nuclear export of HSP70 mRNA. May be involved in the differentiation of granulocytes and adipocytes.Specifications
THOC5 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
C22orf19, Fmip, PK1.3, fSAP79, Fms-interacting protein, NF2/meningioma region protein pK1.3, PP39.2, THO complex subunit 5 homolog, functional spliceosome-associated protein 79, hTREX90, placental protein 39.2 | |
Between 25 and 75 | |
Primary | |
4° C | |
THOC5 |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q13769, Q8BKT7 | |
107829, 8563 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.