Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THOC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543172
Description
Peptide sequence: AGGDCQLHTM DLETGTFTRV LRGHTDYIHC LALRERSPEV LSGGEDGAVR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 79%.
THOC6 belongs to the WD repeat THOC6 family. It contains 7 WD repeats. The function of the THOC6 protein remains unknown.
Specifications
| THOC6 | |
| Polyclonal | |
| Unconjugated | |
| THOC6 | |
| BBIS; F830014G06Rik; fSAP35; Functional spliceosome-associated protein 35; hypothetical protein LOC450075; Pdrp; PSEC0006; THO complex 6; THO complex 6 homolog; THO complex 6 homolog (Drosophila); THO complex subunit 6 homolog; thoc6; WD repeat domain 58; WD repeat-containing protein 58; wdr58; zgc:101618 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79228 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86W42 | |
| THOC6 | |
| Synthetic peptide directed towards the middle region of human THOC6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction