Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THOC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598560 Supplier Invitrogen™ Supplier No. PA598560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

THOC6 Polyclonal Antibody for Western Blot, ELISA

THOC6 belongs to the WD repeat THOC6 family. It contains 7 WD repeats. The function of the THOC6 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THOC6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene THOC6
Gene Accession No. Q86W42
Gene Alias BBIS; F830014G06Rik; fSAP35; Functional spliceosome-associated protein 35; hypothetical protein LOC450075; Pdrp; PSEC0006; THO complex 6; THO complex 6 homolog; THO complex 6 homolog (Drosophila); THO complex subunit 6 homolog; thoc6; WD repeat domain 58; WD repeat-containing protein 58; wdr58; zgc:101618
Gene Symbols THOC6
Host Species Rabbit
Immunogen Recombinant Human THO complex subunit 6 homolog protein (56-315AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79228
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.