Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THOC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545977 Supplier Invitrogen™ Supplier No. PA545977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ELEHLSHIKE SVEDKLELRR KQFHVLLSTI HELQQTLEND EKLSEVEEAQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 100%; Zebrafish: 90%.

THOC7 gene ontology annotations related to this gene include mRNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THOC7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene THOC7
Gene Accession No. Q6I9Y2
Gene Alias FLJ23445; fSAP24; Functional spliceosome-associated protein 24; hTREX30; hypothetical protein LOC445035; im:7153996; Ngg1 interacting factor 3 like 1 binding protein 1; ngg1-interacting factor 3-like protein 1-binding protein 1; NIF3L1-binding protein 1; Nif3l1bp1; RGD1305475; THO complex 7; THO complex 7 homolog; THO complex 7 homolog (Drosophila); THO complex subunit 7 homolog; THO complex subunit 7-like protein; THOC7; THOC7 protein; wu:fa20e08; wu:fj35e07; zgc:92711
Gene Symbols THOC7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human THOC7 (aa 145-194).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80145
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.