Learn More
THOP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305433AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Involved in the metabolism of neuropeptides under 20 amino acid residues long. Involved in cytoplasmic peptide degradation. Able to degrade the beta-amyloid precursor protein and generate amyloidogenic fragments.Specifications
THOP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P52888, Q8C1A5 | |
THOP1 | |
Between 25 and 75 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
THOP1 | |
EP24.15, MEPD_HUMAN, MP78, TOP, endopeptidase 24.15, thimet oligopeptidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50492, 7064 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.