Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ THP-1 TO901317 Treated/ Untreated Cell Lysate
SDP

Catalog No. NB151155 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151155 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151155 Supplier R&D Systems™ Supplier No. NBP311828
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. THP whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 ul per lane for a standard mini-gel blot (approx. 1 mg/ml).

Specifications

Quantity 2 Vials
Storage Buffer The protein lysate is prepared in 1 x Laemmli sample buffer with BME. Boiling before loading is not necessary.
Concentration 1.0 mg/ml
Product Line Novus
Target THP-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.