Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRA Mouse anti-Human, Clone: 2103, Abnova™

Catalog No. 89035576 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-576 Supplier Abnova Corporation Supplier No. MAB14178

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic peptide of human THRA.

  • High specificity
  • High reactivity

Specifications

Antigen THRA
Applications Western Blot
Classification Monoclonal
Clone 2103
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human THRA.
Dilution Western Blot The optimal working dilution should be determined by the end user.
Formulation In HEPES, 0.15 M NaCl, pH 7.5 (50% glycerol, 100 ug/mL BSA).
Gene Accession No. P10827
Gene Alias AR7/EAR7/ERB-T-1/ERBA/ERBA1/MGC000261/MGC43240/NR1A1/THRA1/THRA2/c-ERBA-1
Gene Symbols THRA
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of human THRA.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7067
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.