Learn More
THRA Polyclonal Antibody, Invitrogen™
Description
Recommended positive controls: Colon carcinoma.
Specifications
Specifications
| Antigen | THRA |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | Thra |
| Gene Accession No. | O97716, P10827, P63059 |
| Gene Alias | 6430529J03Rik; AR7; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; AW259572; cerbA protein; c-erb-A protein; c-erb-A thyroid hormone receptor; c-ERBA-1; cerbAalpha; c-erbAalpha; c-erbA-alpha; c-erbAalpha2; c-erbA-alpha2; c-erbAalpha2; thyroid hormone receptor alpha; CHNG6; E ERB-T-1; EAR7; EAR-7; ERBA; ERBA ALPHA; ERBA related 7; ERBA1; ERBA-related 7; ERB-T-1; MGC000261; MGC43240; NR1A1; Nuclear receptor subfamily 1 group A member 1; Oncogene ERBA; Rvr; T3R[a]; T3Ralpha; Thra; THRA1; THRA2; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor alpha 2; thyroid hormone receptor beta; thyroid hormone receptor, alpha; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR; TR alpha 1; TR alpha 1+2; TR alpha 2; triiodothyronine receptor; unnamed protein product; V-erbA-related protein 7 |
| Gene Symbols | Thra |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.