Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRA Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991926
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1926 100 μg
50-199-1927 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1926 Supplier Rockland Immunochemicals Supplier No. 600401A38
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with human THRA protein. A BLAST analysis was used to suggest cross-reactivity with THRA from mouse, human and rat based on a 100% homology with the immunizing sequence. Cross-reactivity with THRA from other sources has not been determined.

The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen THRA
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Thra
Gene Accession No. P10827
Gene Alias 6430529J03Rik; AR7; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; AW259572; cerbA protein; c-erb-A protein; c-erb-A thyroid hormone receptor; c-erbA-1; cerbAalpha; c-erbAalpha; C-erba-alpha; c-erbAalpha2; c-erbA-alpha2; c-erbAalpha2; thyroid hormone receptor alpha; CHNG6; E ERB-T-1; EAR7; EAR-7; ERBA; ERBA ALPHA; ERBA related 7; ERBA1; ERBA-related 7; ERB-T-1; MGC000261; MGC43240; Nr1a1; Nuclear receptor subfamily 1 group A member 1; Oncogene ERBA; Rvr; T3R[a]; T3Ralpha; THRA; THRA1; THRA2; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor alpha 2; thyroid hormone receptor, alpha; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR; TR alpha 1; TR alpha 1+2; TR alpha 2; triiodothyronine receptor; V-erbA-relat
Gene Symbols Thra
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7067
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.