Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987133
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7133 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7133 Supplier Bioss Supplier No. BS6221R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen THRA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Thra
Gene Accession No. P10827
Gene Alias 6430529J03Rik; AR7; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; AW259572; cerbA protein; c-erb-A protein; c-erb-A thyroid hormone receptor; c-erbA-1; cerbAalpha; c-erbAalpha; C-erba-alpha; c-erbAalpha2; c-erbA-alpha2; c-erbAalpha2; thyroid hormone receptor alpha; CHNG6; E ERB-T-1; EAR7; EAR-7; ERBA; ERBA ALPHA; ERBA related 7; ERBA1; ERBA-related 7; ERB-T-1; MGC000261; MGC43240; Nr1a1; Nuclear receptor subfamily 1 group A member 1; Oncogene ERBA; Rvr; T3R[a]; T3Ralpha; THRA; THRA1; THRA2; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor alpha 2; thyroid hormone receptor, alpha; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR; TR alpha 1; TR alpha 1+2; TR alpha 2; triiodothyronine receptor; V-erbA-related protein 7
Gene Symbols Thra
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human THR.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7067
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.