Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566631 Supplier Invitrogen™ Supplier No. PA566631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGKRSEGGHRG FVPEKNFRVT AYKAVQEKSS SPPPRKTSES RDKLGAKGDF PTGKSSFSIT REAQVNVRMD SFDEDLARPS GL Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRAP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THRAP3
Gene Accession No. Q9Y2W1
Gene Alias 9330151F09Rik; B230333E16Rik; BCLAF1 and THRAP3 family member 2; BCLAF2; THRAP3; thyroid hormone receptor associated protein 3; thyroid hormone receptor-associated protein 3; Thyroid hormone receptor-associated protein complex 150 kDa component; thyroid hormone receptor-associated protein, 150 kDa subunit; TRAP150
Gene Symbols THRAP3
Host Species Rabbit
Immunogen Recombinant Human THRAP3. Recombinant protein control fragment (Product #RP-107285).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9967
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.