Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101073 Supplier Invitrogen™ Supplier No. PA5101073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MED24.

This gene encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRAP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MED24
Gene Accession No. O75448, Q4V8B3, Q99K74
Gene Alias 911GSE; activator-recruited cofactor 100 kDa component; ARC100; AU040102; AW547152; Cofactor required for Sp1 transcriptional activation subunit 4; cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa; CRSP complex subunit 4; CRSP100; CRSP4; D11Ertd307e; DRIP100; genetic suppressor element 2; Gse2; hTRAP100; KIAA0130; MED24; MED5; Mediator complex subunit 24; mediator of RNA polymerase II transcription subunit 24; mediator of RNA polymerase II transcription, subunit 24 homolog; MGC8748; mTRAP100; peroxisome proliferation activation receptor binding protein 2; Pparb2; Pparbp2; R75526; THRAP4; thyroid hormone receptor associated protein 4; thyroid hormone receptor-associated protein 100 kDa; thyroid hormone receptor-associated protein 4; Thyroid hormone receptor-associated protein complex 100 kDa component; TRAP100; Vitamin D3 receptor-interacting protein complex 100 kDa component; vitamin D3 receptor-interacting protein complex component DRIP100
Gene Symbols MED24
Host Species Rabbit
Immunogen A synthesized peptide derived from human MED24(Accession O75448), corresponding to amino acid residues V814-N864.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23989, 619436, 9862
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.