Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRAP5 Monoclonal Antibody (3B10)
GREENER_CHOICE

Catalog No. PIMA520890
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520890 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520890 Supplier Invitrogen™ Supplier No. MA520890
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MCDLRRPAAG GMMDLAYVCE WEKWSKSTHC PSVPLACAWS CRNLIAFTMD LRSDDQDLTR MIHILDTEHP WDLHSIPSEH HEAITCLEWD QSGSRLLSAD ADGQIKCWSM.

Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRAP5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MED16
Gene Accession No. Q9Y2X0
Gene Alias 95kDa; A630083L04; DRIP92; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 16; MED16; Mediator complex subunit 16; mediator of RNA polymerase II transcription subunit 16; THRAP5; thyroid hormone receptor associated protein 5; thyroid hormone receptor-associated protein 5; thyroid hormone receptor-associated protein 95; thyroid hormone receptor-associated protein complex 95 kDa component; thyroid hormone receptor-associated protein, 95-kD subunit; TRAP95; Vitamin D3 receptor-interacting protein complex 92 kDa component
Gene Symbols MED16
Host Species Mouse
Immunogen MED16 (AAH17282, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10025
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.