Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1213A
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1213A 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1213A Supplier Invitrogen™ Supplier No. PA1213A
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-213A detects thyroid hormone receptor (TR) beta-1 from human and rat tissues. This antibody does not detect TR alpha-1 or TRv alpha-2. PA1-213A has been successfully used in Western blot, immunohistochemistry (paraffin), IF and gel shift procedures. By Western blot, this antibody detects a ~55 kDa protein representing recombinant human TRv beta-1 expressed in E. coli. PA1-213A immunizing peptide corresponds to residues 62-81 of human TR beta-1, with an N-terminal added cysteine. Alternative numbering of this protein has this peptide corresponding to residues 67-86 of human TR beta-1. This sequence is completely conserved between human and rat TR beta-1.

Thyroid hormone receptors (TR), also known as c-erbA-2, are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences called hormone response elements following activation by the appropriate hormone. Thyroid hormones, through their interaction with TR, effect metabolic processes, growth and development in many tissues by regulating the expression of genes for growth hormone, malic enzyme and several hepatic proteins. There are two forms of TR; TR alpha (on chromosome 17) and TR beta (on chromosome 3). Each of these isoforms also has two isoforms; TR alpha-1 and TRv alpha-2, and TR beta-1 and TR beta-2 respectively. TR alpha-1 and 2 are identical through amino acid 370 where their sequences diverge. TRv alpha-2, which does not bind T3 and is a strong negative regulator of the functional TR, is 80 amino acids longer than TR alpha-1. The corresponding gene for the thyroid hormone receptor beta-1 is NR1A2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRB
Applications Gel Shift, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum, PBS with 0.05% sodium azide
Gene THRB
Gene Accession No. P10828, P18113
Gene Alias c-erbA-2; c-erbAbeta; C-ERBA-BETA; ERBA 2; ERBA BETA; ERBA2; ERBA-BETA; fj24f03; GRTH; LOW QUALITY PROTEIN: thyroid hormone receptor beta; MGC126109; MGC126110; NR1A2; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; PRTH; RATT3REC; si:ch211-264a6.2; T3R[b]; T3Rbeta; T3rec; THR; THR1; thr2; Thrb; Thrb1; Thrb2; thyroid hormone nuclear receptor beta variant 1; thyroid hormone receptor beta; thyroid hormone receptor beta 1; thyroid hormone receptor beta 2; thyroid hormone receptor beta 3; thyroid hormone receptor beta1; thyroid hormone receptor beta-1; thyroid hormone receptor beta2; thyroid hormone receptor beta2delta; thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2); thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); TR; TR beta; TR beta 1; TR[b]1; trb; TR-B1C-ERBA-2; TRbeta; TR-beta; TRbeta1; TRbeta2Delta; wu:fj24f03
Gene Symbols THRB
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C-I(62) F H L D H D D V N D Q S V S S A Q T F(81) of human TR beta-1, with an N-terminal added cysteine.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24831, 7068
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.