Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRB Recombinant Rabbit Monoclonal Antibody (6K1F1)
GREENER_CHOICE

Catalog No. FEMA551306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551306 Supplier Invitrogen™ Supplier No. MA551306
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MTPNSMTENG LTAWDKPKHC PDREHDWKLV GMSEACLHRK SHSERRSTLK NEQSSPHLIQ TTWTSSIFHL DHDDVNDQSV SSAQTFQTEE K.

Thyroid hormone receptors (TR), also known as c-erbA-2, are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences called hormone response elements following activation by the appropriate hormone. Thyroid hormones, through their interaction with TR, effect metabolic processes, growth and development in many tissues by regulating the expression of genes for growth hormone, malic enzyme and several hepatic proteins. There are two forms of TR; TR alpha (on chromosome 17) and TR beta (on chromosome 3). Each of these isoforms also has two isoforms; TR alpha-1 and TRv alpha-2, and TR beta-1 and TR beta-2 respectively. TR alpha-1 and 2 are identical through amino acid 370 where their sequences diverge. TRv alpha-2, which does not bind T3 and is a strong negative regulator of the functional TR, is 80 amino acids longer than TR alpha-1. The corresponding gene for the thyroid hormone receptor beta-1 is NR1A2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRB
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6K1F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene THRB
Gene Accession No. P10828
Gene Alias c-erbA-2; c-erbAbeta; C-ERBA-BETA; ERBA 2; ERBA BETA; ERBA2; ERBA-BETA; fj24f03; GRTH; LOW QUALITY PROTEIN: thyroid hormone receptor beta; MGC126109; MGC126110; NR1A2; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; PRTH; RATT3REC; si:ch211-264a6.2; T3R[b]; T3Rbeta; T3rec; THR; THR1; thr2; Thrb; Thrb1; Thrb2; thyroid hormone nuclear receptor beta variant 1; thyroid hormone receptor beta; thyroid hormone receptor beta 1; thyroid hormone receptor beta 2; thyroid hormone receptor beta 3; thyroid hormone receptor beta1; thyroid hormone receptor beta-1; thyroid hormone receptor beta2; thyroid hormone receptor beta2delta; thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2); thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); TR; TR beta; TR beta 1; TR[b]1; trb; TR-B1C-ERBA-2; TRbeta; TR-beta; TRbeta1; TRbeta2Delta; wu:fj24f03
Gene Symbols THRB
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7068
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.