Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombin-antithrombin (TAT) complexes Antibody (22), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241381AF405
Description
Thrombin-antithrombin (TAT) complexes Monoclonal antibody specifically detects Thrombin-antithrombin (TAT) complexes in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
Thrombin-antithrombin (TAT) complexes | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
F2 | |
Thrombin-antithrombin (TAT) complexes | |
0.1 mL | |
Primary | |
Antibody is specific for human Thrombin-antithrombin (TAT) complexes. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry, ELISA | |
22 | |
Western Blot, Immunohistochemistry, Sandwich ELISA | |
Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin receptor-like 2 | |
Mouse | |
Protein A or G purified | |
Apoptosis, Breast Cancer, Cancer, GPCR, Signal Transduction | |
2147 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction