Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombin-antithrombin (TAT) complexes Antibody (22), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241381MFV610
Description
Thrombin-antithrombin (TAT) complexes Monoclonal antibody specifically detects Thrombin-antithrombin (TAT) complexes in Human samples. It is validated for ImmunohistochemistrySpecifications
| Thrombin-antithrombin (TAT) complexes | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Thrombin-antithrombin (TAT) complexes | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Signal Transduction | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 22 | |
| Immunohistochemistry | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction