Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombin receptor Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0828R

Description
Thrombin is a serine protease that is involved in platelet aggregation and blood coagulation. It is cleaved from its precursor, prothrombin, and converts fibrinogen to fibrin in the final step of the clotting cascade. Thrombin mediates its regulatory effects by activating cell surface G-protein coupled receptors which include PAR-1, PAR-2 and PAR3.Specifications
Thrombin receptor | |
Polyclonal | |
Unconjugated | |
F2R | |
AI482343; C HTR; CF2R; coagulation factor II (thrombin) receptor; Coagulation factor II receptor; coagulation factor II thrombin receptor; F2r; HTR; Par1; PAR-1; protease activated receptor 1; protease-activated receptor 1; Proteinase-activated receptor 1; Thrombin receptor; ThrR; TR; TRGPC | |
Rabbit | |
Protein A | |
RUO | |
2149 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P25116 | |
F2R | |
KLH conjugated synthetic peptide derived from human PAR1 . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction