Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (6C8.2F9), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242213B
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Thrombomodulin/BDCA-3 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
Mouse | |
60.3 kDa | |
0.1 mL | |
Cardiovascular Biology | |
7056 | |
Store at 4C in the dark. | |
IgG1 λ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
6C8.2F9 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P07204 | |
THBD | |
Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204]. | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction