Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (6C8.2F9), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242213MFV500
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Thrombomodulin/BDCA-3 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
6C8.2F9 | |
Flow Cytometry | |
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204]. | |
0.1 mL | |
Cardiovascular Biology | |
7056 | |
Store at 4C in the dark. | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction