missing translation for 'onlineSavingsMsg'
Learn More

Thrombomodulin/BDCA-3 Antibody (6C8.2F9), PE, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242213PE

Numéro de catalogue. NBP242213PE


Il en reste null
ajouter au panier

Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)

Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Flow Cytometry.
Spécifications

Spécifications

Thrombomodulin/BDCA-3
Monoclonal
PE
PBS with 0.05% Sodium Azide
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Mouse
60.3 kDa
0.1 mL
Cardiovascular Biology
7056
Store at 4C in the dark.
IgG1 λ
Flow Cytometry
6C8.2F9
Flow Cytometry
P07204
THBD
Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204].
Protein G purified
RUO
Primary
Human, Mouse
Purified
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Product Certifications
Promotions

Promotions

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'

For Research Use Only