Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Antibody (6C8.2F9), PerCP, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242213PCP

Catalog No. NBP242213P


Only null left
Add to Cart

Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)

Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Thrombomodulin/BDCA-3
Monoclonal
PerCP
PBS with 0.05% Sodium Azide
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Mouse
60.3 kDa
0.1 mL
Cardiovascular Biology
7056
Store at 4C in the dark.
IgG1 λ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
6C8.2F9
Flow Cytometry
P07204
THBD
Partial recombinant human CD141 protein (between amino acids 200-400) [UniProt P07204].
Protein G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only