Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (SR1034), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32253350UL
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
Thrombomodulin/BDCA-3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry | |
SR1034 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
A synthesized peptide derived from human Thrombomodulin/BDCA-3 (Uniprot #: P15306) | |
50 μg | |
Cardiovascular Biology | |
7056 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction